siRNA Details
| Gene Symbol: Galp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022633.1 | NP_072155.1 | 3 | 234 | ||
| GenBank mRNA | AF188491.1 | - | 3 | 234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galp| Gene Symbol: Galp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022633.1 | NP_072155.1 | 3 | 234 | ||
| GenBank mRNA | AF188491.1 | - | 3 | 234 | ||