siRNA Details
| Gene Symbol: Lst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022634.2 | NP_072156.2 | 5 | 302 | ||
| XM_006256078.2 | XP_006256140.1 | 5 | 443 | |||
| XM_006256079.2 | XP_006256141.1 | 5 | 443 | |||
| XM_006256080.2 | XP_006256142.1 | 4 | 390 | |||
| GenBank mRNA | AF208230.1 | - | 5 | 302 | ||
| FQ222284.1 | - | 4 | 300 | |||

Lst1
Genomic Map