siRNA Details
| Gene Symbol: Vax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022637.1 | NP_072159.1 | 3 | 834 | ||
| GenBank mRNA | AF113516.1 | - | 3 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vax2| Gene Symbol: Vax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022637.1 | NP_072159.1 | 3 | 834 | ||
| GenBank mRNA | AF113516.1 | - | 3 | 834 | ||