siRNA Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022684.1 | NP_073175.1 | 3 | 218 | ||
| GenBank mRNA | AF136282.1 | - | 1 | 76 | ||
| AF259503.1 | - | 3 | 218 | |||
| D89804.1 | - | 174 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bid| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022684.1 | NP_073175.1 | 3 | 218 | ||
| GenBank mRNA | AF136282.1 | - | 1 | 76 | ||
| AF259503.1 | - | 3 | 218 | |||
| D89804.1 | - | 174 | ||||