siRNA Details
| Gene Symbol: Snap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022689.2 | NP_073180.1 | 6 | 488 | ||
| GenBank mRNA | AF052596.1 | - | 340 | |||
| BC127494.1 | - | 6 | 488 | 8361815 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snap23| Gene Symbol: Snap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022689.2 | NP_073180.1 | 6 | 488 | ||
| GenBank mRNA | AF052596.1 | - | 340 | |||
| BC127494.1 | - | 6 | 488 | 8361815 | ||