siRNA Details
| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022692.1 | NP_073183.1 | 358 | |||
| GenBank mRNA | AF072935.1 | - | 358 | |||
| BC161848.1 | - | 684 | 9025210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab5a| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022692.1 | NP_073183.1 | 358 | |||
| GenBank mRNA | AF072935.1 | - | 358 | |||
| BC161848.1 | - | 684 | 9025210 | |||