siRNA Details
| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022697.2 | NP_073188.2 | 4 | 1110 | ||
| GenBank mRNA | BC081800.1 | - | 4 | 1110 | 7131175 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl28| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022697.2 | NP_073188.2 | 4 | 1110 | ||
| GenBank mRNA | BC081800.1 | - | 4 | 1110 | 7131175 | |