siRNA Details
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204193.1 | NP_001191122.1 | 5 | 547 | ||
NM_001204194.1 | NP_001191123.1 | 4 | 443 | |||
NM_001204196.1 | NP_001191125.1 | 4 | 439 | |||
NM_003135.2 | NP_003126.1 | 5 | 515 | |||
GenBank mRNA | AK311803.1 | - | 5 | 423 | ||
BC000186.1 | - | 5 | 407 | |||
BC010947.1 | - | 5 | 399 | 4285753 | ||
BC017830.1 | - | 5 | 430 | 4252554 | ||
BU661702.1 | - | 4 | 317 | |||
DN915735.1 | - | 4 | 308 | |||
X12791.1 | - | 5 | 407 |
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BE047584.1 | - | 8 | 540 |
