siRNA Details
| Gene Symbol: Taok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022702.1 | NP_073193.1 | 11 | 1113 | ||
| XM_006230329.2 | XP_006230391.1 | 11 | 2536 | |||
| XM_006230330.2 | XP_006230392.1 | 10 | 957 | |||
| XM_006230331.2 | XP_006230393.1 | 11 | 1824 | |||
| XM_006230332.2 | XP_006230394.1 | 11 | 1825 | |||
| XM_006230334.2 | XP_006230396.1 | 11 | 1809 | |||
| GenBank mRNA | AB290408.1 | - | 10 | 921 | ||
| AF140556.2 | - | 11 | 1113 | |||

Taok2
Genomic Map