siRNA Details
| Gene Symbol: Sgta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022703.3 | NP_073194.1 | 5 | 441 | ||
| XM_006240996.1 | XP_006241058.1 | 5 | 546 | |||
| XM_006240997.1 | XP_006241059.1 | 5 | 543 | |||
| XM_006240998.2 | XP_006241060.1 | 5 | 2386 | |||
| GenBank mRNA | AF368278.1 | - | 5 | 453 | ||
| AJ222724.1 | - | 5 | 429 | |||
| BC087642.1 | - | 5 | 441 | 7308544 | ||

Sgta
Genomic Map