siRNA Details
| Gene Symbol: Pip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022708.2 | NP_073199.1 | 4 | 416 | ||
| GenBank mRNA | AF054270.1 | - | 4 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip| Gene Symbol: Pip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022708.2 | NP_073199.1 | 4 | 416 | ||
| GenBank mRNA | AF054270.1 | - | 4 | 416 | ||