siRNA Details
| Gene Symbol: SRP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003136.3 | NP_003127.1 | 2, 3 | 432 | ||
| XM_005268024.2 | XP_005268081.1 | 2, 3 | 685 | |||
| XM_011537106.1 | XP_011535408.1 | 2, 3 | 313 | |||
| XM_017021615.1 | XP_016877104.1 | 2, 3 | 357 | |||
| GenBank mRNA | AK312853.1 | - | 2, 3 | 377 | ||
| BC000652.2 | - | 2, 3 | 299 | 3343314 | ||
| BC003389.1 | - | 2, 3 | 305 | 3446599 | ||
| U51920.1 | - | 2, 3 | 152 | |||

SRP54
Genomic Map