siRNA Details
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022715.2 | NP_073206.2 | 14 | 2342 | ||
| GenBank mRNA | BC071174.1 | - | 14 | 2349 | 7099451 | |
| U09870.2 | - | 14 | 2342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mvp| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022715.2 | NP_073206.2 | 14 | 2342 | ||
| GenBank mRNA | BC071174.1 | - | 14 | 2349 | 7099451 | |
| U09870.2 | - | 14 | 2342 | |||