siRNA Details
| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022800.1 | NP_073637.1 | 2 | 556 | ||
| GenBank mRNA | AF313450.1 | - | 2 | 556 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2ry12| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022800.1 | NP_073637.1 | 2 | 556 | ||
| GenBank mRNA | AF313450.1 | - | 2 | 556 | ||
Set Membership: |
|