siRNA Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022855.2 | NP_074046.1 | 8 | 1700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g3| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022855.2 | NP_074046.1 | 8 | 1700 | ||