siRNA Details
| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022861.1 | NP_074052.1 | 3296 | |||
| GenBank mRNA | U24070.1 | - | 3296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13a| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022861.1 | NP_074052.1 | 3296 | |||
| GenBank mRNA | U24070.1 | - | 3296 | |||