siRNA Details
| Gene Symbol: Unc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042579.1 | NP_001036044.1 | 43 | 4874 | ||
| NM_022862.1 | NP_074053.1 | 34 | 6348 | |||
| GenBank mRNA | AF159706.1 | - | 43 | 4874 | ||
| U24071.1 | - | 34 | 6348 | |||

Unc13b
Genomic Map