siRNA Details
| Gene Symbol: Cplx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022864.3 | NP_074055.1 | 2 | 243 | ||
| XM_006250571.2 | XP_006250633.1 | 3 | 369 | |||
| GenBank mRNA | BC093605.1 | - | 2 | 246 | 7317504 | |
| D70817.1 | - | 3 | 278 | |||
| U35098.1 | - | 3 | 276 | |||

Cplx1
Genomic Map