siRNA Details
| Gene Symbol: Aoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022935.2 | NP_075224.2 | 2 | 1150 | ||
| XM_006236471.2 | XP_006236533.1 | 2 | 1172 | |||
| XM_006236472.2 | XP_006236534.1 | 2 | 993 | |||
| XM_008762920.1 | XP_008761142.1 | 2 | 1174 | |||
| GenBank mRNA | BC100144.1 | - | 2 | 1150 | 7460184 | |
| X73911.1 | - | 2 | 1032 | |||

Aoc1
Genomic Map