siRNA Details
| Gene Symbol: Htr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022938.2 | NP_075227.1 | 2 | 1259 | ||
| XM_006231307.2 | XP_006231369.1 | 2 | 1474 | |||
| GenBank mRNA | L22558.1 | - | 2 | 1259 | ||
| U68489.1 | - | 2 | 1259 | |||
| U68490.1 | - | 3 | 1259 | |||
| X69663.1 | - | 2 | 1442 | |||
| Gene Symbol: LOC103694905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773900.1 | XP_008772122.1 | 1474 | |||
| XM_008773901.1 | XP_008772123.1 | 1474 | ||||
More Information
Set Membership: |
|

Htr7
Genomic Map