siRNA Details
| Gene Symbol: Stx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022939.2 | NP_075228.2 | 9 | 862 | ||
| GenBank mRNA | AF035632.1 | - | 9 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx12| Gene Symbol: Stx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022939.2 | NP_075228.2 | 9 | 862 | ||
| GenBank mRNA | AF035632.1 | - | 9 | 778 | ||