siRNA Details
| Gene Symbol: Pdzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022940.1 | NP_075229.1 | 4413 | |||
| GenBank mRNA | AF169411.1 | - | 4413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzd2| Gene Symbol: Pdzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022940.1 | NP_075229.1 | 4413 | |||
| GenBank mRNA | AF169411.1 | - | 4413 | |||