siRNA Details
| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022943.1 | NP_075232.1 | 19 | 2939 | ||
| GenBank mRNA | AF208235.1 | - | 19 | 2939 | ||
| FQ221127.1 | - | 2585 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mertk| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022943.1 | NP_075232.1 | 19 | 2939 | ||
| GenBank mRNA | AF208235.1 | - | 19 | 2939 | ||
| FQ221127.1 | - | 2585 | ||||