siRNA Details
| Gene Symbol: Sfxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022948.1 | NP_075237.1 | 7 | 848 | ||
| XM_006231563.2 | XP_006231625.1 | 8 | 998 | |||
| XM_006231565.2 | XP_006231627.1 | 7 | 716 | |||
| XM_006231566.2 | XP_006231628.1 | 7 | 738 | |||
| XM_008760451.1 | XP_008758673.1 | 7 | 735 | |||
| GenBank mRNA | AF276997.1 | - | 7 | 848 | ||
| BC062043.1 | - | 7 | 877 | 5597953 | ||

Sfxn3
Genomic Map