siRNA Details
| Gene Symbol: C1galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022950.1 | NP_075239.1 | 2 | 284 | ||
| XM_006236091.2 | XP_006236153.1 | 3 | 305 | |||
| XM_008762725.1 | XP_008760947.1 | 3 | 254 | |||
| GenBank mRNA | AF157963.1 | - | 2 | 284 | ||

C1galt1
Genomic Map