siRNA Details
| Gene Symbol: Ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022952.2 | NP_075241.2 | 5 | 410 | ||
| GenBank mRNA | BC088138.1 | - | 5 | 410 | 7374432 | |
| FQ234449.1 | - | 5 | 429 | |||
| M37194.1 | - | 5 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap2s1| Gene Symbol: Ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022952.2 | NP_075241.2 | 5 | 410 | ||
| GenBank mRNA | BC088138.1 | - | 5 | 410 | 7374432 | |
| FQ234449.1 | - | 5 | 429 | |||
| M37194.1 | - | 5 | 412 | |||