siRNA Details
| Gene Symbol: Slit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022953.2 | NP_075242.1 | 10 | 1115 | ||
| GenBank mRNA | AB011530.1 | - | 10 | 1115 | ||
| AB073213.1 | - | 9 | 962 | |||
| AB073214.1 | - | 10 | 1040 | |||
| AB073215.1 | - | 10 | 1040 | |||
| AF133730.1 | - | 10 | 956 | |||

Slit1
Genomic Map