siRNA Details
| Gene Symbol: Serpina5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244739.1 | NP_001231668.1 | 3 | 826 | ||
| NM_022957.3 | NP_075246.3 | 3 | 884 | |||
| XM_006240494.2 | XP_006240556.1 | 6 | 2999 | |||
| GenBank mRNA | AB013128.1 | - | 3 | 757 | ||
| BC081797.1 | - | 3 | 794 | 7112069 | ||
| BC097315.1 | - | 3 | 841 | 7453034 | ||

Serpina5
Genomic Map