siRNA Details
| Gene Symbol: Aqp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022960.2 | NP_075249.1 | 6 | 638 | ||
| XM_006243362.1 | XP_006243424.1 | 5 | 743 | |||
| XM_008766336.1 | XP_008764558.1 | 2 | 444 | |||
| GenBank mRNA | AF016406.1 | - | 2 | 417 | ||
| BC085731.1 | - | 6 | 638 | 7134715 | ||
| FQ209455.1 | - | 2 | 439 | |||
| FQ211101.1 | - | 2 | 359 | |||
| FQ212527.1 | - | 2 | 322 | |||
| FQ214432.1 | - | 5 | 748 | |||
| FQ218988.1 | - | 2 | 442 | |||
| FQ219175.1 | - | 2 | 359 | |||

Aqp9
Genomic Map