siRNA Details
| Gene Symbol: Aqp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022960.2 | NP_075249.1 | 10 | 1254 | ||
| XM_006243362.1 | XP_006243424.1 | 9 | 1359 | |||
| XM_008766336.1 | XP_008764558.1 | 6 | 1060 | |||
| GenBank mRNA | AF016406.1 | - | 6 | 1033 | ||
| BC085731.1 | - | 10 | 1254 | 7134715 | ||
| FQ209455.1 | - | 6 | 1056 | |||
| FQ212527.1 | - | 6 | 939 | |||
| FQ214432.1 | - | 9 | 1364 | |||
| FQ218988.1 | - | 6 | 1058 | |||
| FQ219175.1 | - | 6 | 975 | |||

Aqp9
Genomic Map