siRNA Details
| Gene Symbol: Vps33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022961.3 | NP_075250.2 | 10 | 1445 | ||
| GenBank mRNA | BC105752.1 | - | 10 | 1445 | 5599123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps33a| Gene Symbol: Vps33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022961.3 | NP_075250.2 | 10 | 1445 | ||
| GenBank mRNA | BC105752.1 | - | 10 | 1445 | 5599123 | |