siRNA Details
| Gene Symbol: Cldn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031699.2 | NP_113887.2 | 3 | 609 | ||
| GenBank mRNA | AF195500.1 | - | 3 | 550 | ||
| BC061992.1 | - | 3 | 609 | 5599462 | ||
| FQ220868.1 | - | 3 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn1| Gene Symbol: Cldn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031699.2 | NP_113887.2 | 3 | 609 | ||
| GenBank mRNA | AF195500.1 | - | 3 | 550 | ||
| BC061992.1 | - | 3 | 609 | 5599462 | ||
| FQ220868.1 | - | 3 | 578 | |||