siRNA Details
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031702.1 | NP_113890.1 | 4, 5 | 778 | ||
XM_006246805.2 | XP_006246867.1 | 3, 4 | 978 | |||
GenBank mRNA | AJ011811.1 | - | 3, 4 | 414 | ||
BC101892.1 | - | 4, 5 | 778 | 7930797 | ||
BC159404.1 | - | 3, 4 | 872 | 7458165 |
