siRNA Details
| Gene Symbol: Pdzk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031712.1 | NP_113900.1 | 8 | 1459 | ||
| GenBank mRNA | AF013145.1 | - | 8 | 1430 | ||
| AF116896.1 | - | 8 | 1459 | |||
| BC078788.1 | - | 8 | 1451 | 7109724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzk1| Gene Symbol: Pdzk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031712.1 | NP_113900.1 | 8 | 1459 | ||
| GenBank mRNA | AF013145.1 | - | 8 | 1430 | ||
| AF116896.1 | - | 8 | 1459 | |||
| BC078788.1 | - | 8 | 1451 | 7109724 | ||