siRNA Details
| Gene Symbol: Pfkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031715.1 | NP_113903.1 | 594 | |||
| GenBank mRNA | BC094212.1 | - | 5 | 581 | 7314667 | |
| D21869.1 | - | 5 | 538 | |||
| FN433010.1 | - | 5 | 559 | |||
| FQ211719.1 | - | 720 | ||||
| U25651.1 | - | 594 | ||||

Pfkm
Genomic Map