siRNA Details
| Gene Symbol: Ncs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024366.1 | NP_077342.1 | 5 | 404 | ||
| XM_006233925.2 | XP_006233987.1 | 5 | 708 | |||
| XM_008761679.1 | XP_008759901.1 | 4 | 299 | |||
| GenBank mRNA | L27421.1 | - | 5 | 344 | ||
| X82188.1 | - | 5 | 404 | |||

Ncs1
Genomic Map