siRNA Details
| Gene Symbol: Clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031719.1 | NP_113907.1 | 2 | 255 | ||
| XM_006229792.2 | XP_006229854.1 | 2 | 318 | |||
| GenBank mRNA | BC060555.1 | - | 2 | 299 | 6918578 | |
| D13985.1 | - | 2 | 325 | |||
| D17698.1 | - | 2 | 238 | |||
| L26450.1 | - | 2 | 255 | |||

Clns1a
Genomic Map