siRNA Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031721.1 | NP_113909.1 | 2 | 544 | ||
| GenBank mRNA | AF179370.1 | - | 2 | 544 | ||
| BC081767.1 | - | 2 | 569 | 7121959 | ||
| FQ228942.1 | - | 1 | 22 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htra1| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031721.1 | NP_113909.1 | 2 | 544 | ||
| GenBank mRNA | AF179370.1 | - | 2 | 544 | ||
| BC081767.1 | - | 2 | 569 | 7121959 | ||
| FQ228942.1 | - | 1 | 22 | |||