siRNA Details
| Gene Symbol: Tmed2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031722.2 | NP_113910.1 | 2 | 371 | ||
| XM_006249327.2 | XP_006249389.1 | 2 | 420 | |||
| GenBank mRNA | BC062036.1 | - | 2 | 371 | 5623841 | |
| X92097.1 | - | 2 | 342 | |||

Tmed2
Genomic Map