siRNA Details
| Gene Symbol: Sec11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031723.3 | NP_113911.2 | 4, 5 | 520 | ||
| XM_008759558.1 | XP_008757780.1 | 4, 5 | 476 | |||
| GenBank mRNA | BC060554.1 | - | 4, 5 | 524 | 6921802 | |
| FQ220501.1 | - | 4, 5 | 524 | |||
| FQ221649.1 | - | 2, 3 | 227 | |||
| L11319.1 | - | 4, 5 | 507 | |||

Sec11a
Genomic Map