siRNA Details
| Gene Symbol: Scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023955.1 | NP_076445.1 | 5 | 544 | ||
| GenBank mRNA | AF295405.1 | - | 4 | 388 | ||
| BC085862.1 | - | 5 | 544 | 7190371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scamp2| Gene Symbol: Scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023955.1 | NP_076445.1 | 5 | 544 | ||
| GenBank mRNA | AF295405.1 | - | 4 | 388 | ||
| BC085862.1 | - | 5 | 544 | 7190371 | ||