siRNA Details
| Gene Symbol: Scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031724.1 | NP_113912.1 | 6, 7 | 804 | ||
| XM_006232753.2 | XP_006232815.1 | 6, 7 | 1259 | |||
| XM_006232754.2 | XP_006232816.1 | 6, 7 | 1309 | |||
| GenBank mRNA | AF295404.1 | - | 3, 4 | 323 | ||
| DV726194.1 | - | 3, 4 | 345 | |||

Scamp3
Genomic Map