siRNA Details
| Gene Symbol: Snap91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031728.1 | NP_113916.1 | 8, 9 | 825 | ||
| GenBank mRNA | X68877.1 | - | 8, 9 | 825 | ||
| X68878.1 | - | 8, 9 | 825 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snap91| Gene Symbol: Snap91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031728.1 | NP_113916.1 | 8, 9 | 825 | ||
| GenBank mRNA | X68877.1 | - | 8, 9 | 825 | ||
| X68878.1 | - | 8, 9 | 825 | |||