siRNA Details
| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031730.2 | NP_113918.2 | 4 | 1953 | ||
| XM_008762830.1 | XP_008761052.1 | 4 | 477 | |||
| GenBank mRNA | CO393575.1 | - | 4 | 514 | ||
| M59980.1 | - | 4 | 1632 | |||
| S64320.1 | - | 4 | 1951 | |||

Kcnd2
Genomic Map