siRNA Details
| Gene Symbol: Prol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133513.1 | NP_598197.1 | 3 | 248 | ||
| GenBank mRNA | U03407.1 | - | 3 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prol1| Gene Symbol: Prol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133513.1 | NP_598197.1 | 3 | 248 | ||
| GenBank mRNA | U03407.1 | - | 3 | 248 | ||