siRNA Details
| Gene Symbol: Aldh3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031731.2 | NP_113919.2 | 3, 4 | 597 | ||
| XM_006246529.2 | XP_006246591.1 | 3, 4 | 1095 | |||
| GenBank mRNA | CB727458.1 | - | 1, 2 | 60 | ||
| FQ210485.1 | - | 3, 4 | 660 | |||
| FQ221576.1 | - | 3, 4 | 619 | |||
| M73714.1 | - | 3, 4 | 603 | |||

Aldh3a2
Genomic Map