siRNA Details
Gene Symbol: Gabrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031733.1 | NP_113921.1 | 9 | 1377 | ||
XM_006229516.2 | XP_006229578.1 | 10 | 1541 | |||
GenBank mRNA | AF419333.1 | - | 9 | 1377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031733.1 | NP_113921.1 | 9 | 1377 | ||
XM_006229516.2 | XP_006229578.1 | 10 | 1541 | |||
GenBank mRNA | AF419333.1 | - | 9 | 1377 |