siRNA Details
| Gene Symbol: Slc29a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031738.1 | NP_113926.1 | 4 | 556 | ||
| XM_008760174.1 | XP_008758396.1 | 2309 | ||||
| GenBank mRNA | AF015305.1 | - | 4 | 556 | ||
| Gene Symbol: LOC100911721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749035.3 | XP_003749083.1 | 4 | 1340 | ||
| XM_008760194.1 | XP_008758416.1 | 4 | 2309 | |||

Slc29a2
Genomic Map