siRNA Details
| Gene Symbol: Kcnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270962.1 | NP_001257891.1 | 8 | 2438 | ||
| NM_001270963.1 | NP_001257892.1 | 7 | 2381 | |||
| NM_031739.3 | NP_113927.2 | 8 | 1887 | |||
| GenBank mRNA | AB008805.1 | - | 2 | 141 | ||
| AB008806.1 | - | 2 | 134 | |||
| L48619.1 | - | 8 | 1894 | |||

Kcnd3
Genomic Map