siRNA Details
| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031742.1 | NP_113930.1 | 9 | 1839 | ||
| XM_006250491.2 | XP_006250553.1 | 9 | 1856 | |||
| GenBank mRNA | Z34264.1 | - | 9 | 1839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh1| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031742.1 | NP_113930.1 | 9 | 1839 | ||
| XM_006250491.2 | XP_006250553.1 | 9 | 1856 | |||
| GenBank mRNA | Z34264.1 | - | 9 | 1839 | ||